Assay Details
Gene Symbol: opn1mw4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131254.1 | NP_571329.1 | 1 - 2 | 438 | 73 | |
GenBank mRNA | AF109370.1 | - | 1 - 2 | 438 | 73 | |
BC059503.1 | - | 1 - 2 | 432 | 73 | ||
KT008397.1 | - | 1 - 2 | 360 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
