Assay Details
Gene Symbol: celf3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131494.2 | NP_571569.2 | 13 - 14 | 1577 | 76 | |
XM_021468129.1 | XP_021323804.1 | 13 - 14 | 1673 | 76 | ||
GenBank mRNA | AB032725.1 | - | 12 - 13 | 1401 | 76 | |
BC060923.1 | - | 13 - 14 | 1577 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
