Assay Details
| Gene Symbol: celf3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131494.2 | NP_571569.2 | 4 - 5 | 486 | 59 | |
| XM_021468129.1 | XP_021323804.1 | 4 - 5 | 564 | 59 | ||
| GenBank mRNA | AB032725.1 | - | 3 - 4 | 310 | 59 | |
| BC060923.1 | - | 4 - 5 | 486 | 59 | ||
More Information
Set Membership: |
|

celf3a
Genomic Map