Assay Details
Gene Symbol: ccnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131513.1 | NP_571588.1 | 3 - 4 | 366 | 132 | |
GenBank mRNA | AB040435.1 | - | 3 - 4 | 366 | 132 | |
AF268043.1 | - | 3 - 4 | 380 | 132 | ||
BC045492.1 | - | 3 - 4 | 372 | 132 | ||
BC055553.1 | - | 3 - 4 | 331 | 132 | ||
BC067192.1 | - | 3 - 4 | 360 | 132 | ||
BC153626.1 | - | 3 - 4 | 283 | 132 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
