Assay Details
| Gene Symbol: prss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131708.2 | NP_571783.2 | 1 - 2 | 58 | 90 | |
| GenBank mRNA | AF541952.1 | - | 1 - 2 | 41 | 90 | |
| BC092664.1 | - | 1 - 2 | 55 | 90 | ||
| BC165931.1 | - | 1 - 2 | 68 | 90 | ||
| EH490900.1 | - | 1 - 2 | 88 | 90 | ||
| JQ999998.1 | - | 1 - 2 | 52 | 90 | ||
More Information
Set Membership: |
|

prss1
Genomic Map