Assay Details
| Gene Symbol: psma6l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131795.2 | NP_571870.2 | 6 - 7 | 770 | 64 | |
| GenBank mRNA | AF354751.1 | - | 6 - 7 | 1005 | 64 | |
| BC072719.1 | - | 6 - 7 | 770 | 64 | ||
| BC165117.1 | - | 6 - 7 | 736 | 64 | ||
More Information
Set Membership: |
|

psma6l
Genomic Map