Assay Details
| Gene Symbol: tfe3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131848.2 | NP_571923.2 | 2 - 3 | 617 | 126 | |
| XM_005166704.4 | XP_005166761.1 | 2 - 3 | 381 | 126 | ||
| GenBank mRNA | AF362684.1 | - | 2 - 3 | 448 | 126 | |
| BC092697.1 | - | 2 - 3 | 617 | 126 | ||
| BC165184.1 | - | 2 - 3 | 282 | 126 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005166704.4 transcripts.
Set Membership: |
|

tfe3a
Genomic Map