Assay Details
| Gene Symbol: tfe3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131848.2 | NP_571923.2 | 4 - 5 | 1064 | 98 | |
| XM_005166704.4 | XP_005166761.1 | 4 - 5 | 828 | 98 | ||
| GenBank mRNA | AF362684.1 | - | 4 - 5 | 895 | 98 | |
| BC092697.1 | - | 4 - 5 | 1064 | 98 | ||
| BC165184.1 | - | 4 - 5 | 729 | 98 | ||
More Information
Set Membership: |
|

tfe3a
Genomic Map