Assay Details
| Gene Symbol: tfe3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131848.2 | NP_571923.2 | 8 - 9 | 1417 | 79 | |
| XM_005166704.4 | XP_005166761.1 | 8 - 9 | 1181 | 79 | ||
| XM_005166705.4 | XP_005166762.1 | 4 - 5 | 696 | 79 | ||
| GenBank mRNA | AF362684.1 | - | 8 - 9 | 1248 | 79 | |
| BC092697.1 | - | 8 - 9 | 1417 | 79 | ||
| BC165184.1 | - | 8 - 9 | 1082 | 79 | ||
| JN105111.1 | - | 4 - 5 | 307 | 79 | ||
More Information
Set Membership: |
|

tfe3a
Genomic Map