Assay Details
| Gene Symbol: sox32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131851.1 | NP_571926.1 | 1 - 2 | 328 | 67 | |
| GenBank mRNA | AB071895.1 | - | 1 - 2 | 341 | 67 | |
| AF362749.1 | - | 1 - 2 | 328 | 67 | ||
| AY027650.1 | - | 1 - 2 | 313 | 67 | ||
| BC097087.1 | - | 1 - 2 | 342 | 67 | ||
More Information
Set Membership: |
|

sox32
Genomic Map