Assay Details
Gene Symbol: nrp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181497.3 | NP_852474.2 | 11 - 12 | 1972 | 73 | |
XM_009297197.3 | XP_009295472.1 | 11 - 12 | 2476 | 73 | ||
GenBank mRNA | AB088776.1 | - | 11 - 12 | 1866 | 73 | |
AY064213.1 | - | 11 - 12 | 2176 | 73 | ||
AY351910.1 | - | 11 - 12 | 1972 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
