Assay Details
| Gene Symbol: keap1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182864.2 | NP_878284.2 | 2 - 3 | 899 | 69 | |
| GenBank mRNA | AB081315.2 | - | 2 - 3 | 835 | 69 | |
| BC094989.1 | - | 867 | 69 | |||
| BC117613.1 | - | 2 - 3 | 899 | 69 | ||
More Information
Set Membership: |
|

keap1a
Genomic Map