Assay Details
Gene Symbol: rdh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198069.1 | NP_932335.1 | 2 - 3 | 397 | 57 | |
XM_005167928.3 | XP_005167985.1 | 2 - 3 | 446 | 57 | ||
GenBank mRNA | BC046089.1 | - | 2 - 3 | 397 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
