Assay Details
| Gene Symbol: rdh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198069.1 | NP_932335.1 | 3 - 4 | 649 | 101 | |
| XM_005167928.3 | XP_005167985.1 | 3 - 4 | 101 | |||
| GenBank mRNA | BC046089.1 | - | 3 - 4 | 649 | 101 | |
More Information
Set Membership: |
|

rdh1
Genomic Map