Assay Details
| Gene Symbol: rdh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198069.1 | NP_932335.1 | 4 - 5 | 818 | 103 | |
| GenBank mRNA | BC046089.1 | - | 4 - 5 | 818 | 103 | |
More Information
Set Membership: |
|

rdh1
Genomic Map