Assay Details
| Gene Symbol: dohh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199563.2 | NP_955857.1 | 3 - 4 | 526 | 113 | |
| GenBank mRNA | BC046086.1 | - | 3 - 4 | 527 | 113 | |
More Information
Set Membership: |
|

dohh
Genomic Map