Assay Details
| Gene Symbol: zc2hc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199680.1 | NP_955974.1 | 6 - 7 | 588 | 81 | |
| XM_005162676.4 | XP_005162733.1 | 5 - 6 | 590 | 81 | ||
| GenBank mRNA | BC055250.1 | - | 6 - 7 | 588 | 81 | |
More Information
Set Membership: |
|

zc2hc1a
Genomic Map