Assay Details
| Gene Symbol: ptpn11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199846.1 | NP_956140.1 | 10 - 11 | 1425 | 69 | |
| GenBank mRNA | AF097478.1 | - | 1 - 2 | 133 | 69 | |
| BC045328.1 | - | 10 - 11 | 1425 | 69 | ||
| BC164416.1 | - | 9 - 10 | 1265 | 69 | ||
| FN428738.1 | - | 2 - 3 | 181 | 69 | ||
More Information
Set Membership: |
|

ptpn11a
Genomic Map