Assay Details
| Gene Symbol: ptpn11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199846.1 | NP_956140.1 | 5 - 6 | 840 | 65 | |
| GenBank mRNA | BC045328.1 | - | 5 - 6 | 840 | 65 | |
| BC164416.1 | - | 4 - 5 | 680 | 65 | ||
More Information
Set Membership: |
|

ptpn11a
Genomic Map