Assay Details
| Gene Symbol: porcnl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199849.1 | NP_956143.1 | 10 - 11 | 1119 | 93 | |
| GenBank mRNA | BC044156.1 | - | 10 - 11 | 1119 | 93 | |
| BC154563.1 | - | 10 - 11 | 1230 | 93 | ||
More Information
Set Membership: |
|

porcnl
Genomic Map