Assay Details
| Gene Symbol: zc3h15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199888.1 | NP_956182.1 | 9 - 10 | 1217 | 69 | |
| GenBank mRNA | BC044451.1 | - | 9 - 10 | 1217 | 69 | |
| BC165333.1 | - | 9 - 10 | 1144 | 69 | ||
More Information
Set Membership: |
|

zc3h15
Genomic Map