Assay Details
Gene Symbol: ppiab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328424.1 | NP_001315353.1 | 2 - 3 | 296 | 97 | |
GenBank mRNA | AY391451.1 | - | 2 - 3 | 164 | 97 | |
BC059470.1 | - | 2 - 3 | 241 | 97 | ||
BC071370.1 | - | 2 - 3 | 173 | 97 | ||
CF998539.1 | - | 2 - 3 | 143 | 97 | ||
EH487851.1 | - | 2 - 3 | 233 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
