Assay Details
| Gene Symbol: ppiab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328424.1 | NP_001315353.1 | 4 - 5 | 557 | 67 | |
| GenBank mRNA | BC059470.1 | - | 4 - 5 | 502 | 67 | |
| BC071370.1 | - | 4 - 5 | 434 | 67 | ||
| CF998539.1 | - | 4 - 5 | 404 | 67 | ||
| EH487851.1 | - | 4 - 5 | 494 | 67 | ||
More Information
Set Membership: |
|

ppiab
Genomic Map