Assay Details
| Gene Symbol: hmgcs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201085.2 | NP_957379.2 | 2 - 3 | 548 | 80 | |
| XM_005155491.3 | XP_005155548.1 | 2 - 3 | 592 | 80 | ||
| GenBank mRNA | BC049456.1 | - | 2 - 3 | 545 | 80 | |
More Information
Set Membership: |
|

hmgcs1
Genomic Map