Assay Details
Gene Symbol: mcm3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171068.1 | NP_001164539.1 | 11 - 12 | 1789 | 104 | |
GenBank mRNA | AB973819.1 | - | 11 - 12 | 1670 | 104 | |
BC045431.1 | - | 11 - 12 | 1777 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
