Assay Details
Gene Symbol: mcm3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171068.1 | NP_001164539.1 | 4 - 5 | 645 | 135 | |
GenBank mRNA | BC045431.1 | - | 4 - 5 | 633 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
