Assay Details
| Gene Symbol: dmrt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205628.2 | NP_991191.2 | 1 - 2 | 357 | 73 | |
| NM_205761.2 | NP_991324.2 | 1 - 2 | 357 | 73 | ||
| GenBank mRNA | AB127117.1 | - | 1 - 2 | 149 | 73 | |
| AF439562.1 | - | 1 - 2 | 227 | 73 | ||
| AY157561.1 | - | 1 - 2 | 224 | 73 | ||
| AY157562.1 | - | 1 - 2 | 224 | 73 | ||
| AY157563.1 | - | 1 - 2 | 224 | 73 | ||
| AY157564.1 | - | 1 - 2 | 224 | 73 | ||
| AY157565.1 | - | 1 - 2 | 224 | 73 | ||
| AY157566.1 | - | 1 - 2 | 224 | 73 | ||
| AY157567.1 | - | 1 - 2 | 224 | 73 | ||
| AY157568.1 | - | 1 - 2 | 224 | 73 | ||
| AY157569.1 | - | 1 - 2 | 224 | 73 | ||
| AY157570.1 | - | 1 - 2 | 224 | 73 | ||
| AY157571.1 | - | 1 - 2 | 224 | 73 | ||
| AY566608.1 | - | 1 - 2 | 318 | 73 | ||
| AY588966.1 | - | 1 - 2 | 318 | 73 | ||
| BC116610.1 | - | 1 - 2 | 239 | 73 | ||
More Information
Set Membership: |
|

dmrt1
Genomic Map