Assay Details
| Gene Symbol: lman2la | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205725.2 | NP_991288.2 | 4 | 463 | 80 | |
| GenBank mRNA | BC097031.1 | - | 4 | 463 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

lman2la| Gene Symbol: lman2la | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205725.2 | NP_991288.2 | 4 | 463 | 80 | |
| GenBank mRNA | BC097031.1 | - | 4 | 463 | 80 | |
Set Membership: |
|