Assay Details
Gene Symbol: ptk2bb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212570.1 | NP_997735.1 | 4 - 5 | 633 | 100 | |
GenBank mRNA | AJ490510.2 | - | 4 - 5 | 633 | 100 | |
BC134027.1 | - | 5 - 6 | 664 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
