Assay Details
| Gene Symbol: hm13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212572.2 | NP_997737.2 | 9 - 10 | 990 | 132 | |
| XM_005161890.4 | XP_005161947.1 | 9 - 10 | 1027 | 132 | ||
| GenBank mRNA | AY926458.1 | - | 9 - 10 | 956 | 132 | |
| BC053414.1 | - | 9 - 10 | 927 | 132 | ||
| BC071503.1 | - | 9 - 10 | 990 | 132 | ||
More Information
Set Membership: |
|

hm13
Genomic Map