Assay Details
Gene Symbol: st6galnac3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212625.2 | NP_997790.1 | 3 - 4 | 740 | 71 | |
GenBank mRNA | AJ620947.1 | - | 3 - 4 | 740 | 71 | |
AJ705081.1 | - | 3 - 4 | 626 | 71 | ||
BC162117.1 | - | 3 - 4 | 656 | 71 | ||
BC162495.1 | - | 3 - 4 | 656 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
