Assay Details
| Gene Symbol: alcamb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212634.3 | NP_997799.3 | 3 - 4 | 526 | 85 | |
| XM_005157504.4 | XP_005157561.1 | 3 - 4 | 537 | 85 | ||
| GenBank mRNA | AY050561.1 | - | 3 - 4 | 482 | 85 | |
| BC163458.1 | - | 3 - 4 | 432 | 85 | ||
| BC163486.1 | - | 3 - 4 | 432 | 85 | ||
| CU458904.1 | - | 3 - 4 | 396 | 85 | ||
| CU458936.1 | - | 3 - 4 | 448 | 85 | ||
| DQ279080.1 | - | 3 - 4 | 489 | 85 | ||
More Information
Set Membership: |
|

alcamb
Genomic Map