Assay Details
| Gene Symbol: tgm2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212656.2 | NP_997821.2 | 3 - 4 | 433 | 89 | |
| GenBank mRNA | AF242545.2 | - | 3 - 4 | 456 | 89 | |
| BC117635.1 | - | 3 - 4 | 433 | 89 | ||
More Information
Set Membership: |
|

tgm2b
Genomic Map