Assay Details
| Gene Symbol: glcea | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212850.2 | NP_998015.2 | 3 - 4 | 878 | 91 | |
| XM_017354068.2 | XP_017209557.1 | 3 - 4 | 949 | 91 | ||
| GenBank mRNA | AY388516.1 | - | 2 - 3 | 733 | 91 | |
| BC114251.1 | - | 3 - 4 | 878 | 91 | ||
More Information
Set Membership: |
|

glcea
Genomic Map