Assay Details
Gene Symbol: pak5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212962.1 | NP_998127.1 | 6 - 7 | 2115 | 78 | |
GenBank mRNA | BC068016.1 | - | 6 - 7 | 2115 | 78 | |
BC168506.1 | - | 6 - 7 | 2010 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
