Assay Details
Gene Symbol: ptges3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213170.1 | NP_998335.1 | 6 - 7 | 605 | 117 | |
GenBank mRNA | BC056294.1 | - | 6 - 7 | 596 | 117 | |
BC065314.1 | - | 6 - 7 | 605 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
