Assay Details
| Gene Symbol: ak4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213299.1 | NP_998464.1 | 3 - 4 | 347 | 68 | |
| XM_005165871.3 | XP_005165928.1 | 4 - 5 | 891 | 68 | ||
| XM_005165873.3 | XP_005165930.1 | 4 - 5 | 887 | 68 | ||
| GenBank mRNA | BC068387.1 | - | 3 - 4 | 347 | 68 | |
More Information
Set Membership: |
|

ak4
Genomic Map