Assay Details
| Gene Symbol: gab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001326713.1 | NP_001313642.1 | 11 - 12 | 2606 | 66 | |
| XM_009292494.3 | XP_009290769.1 | 10 - 11 | 2588 | 66 | ||
| XM_017353102.2 | XP_017208591.1 | 12 - 13 | 2766 | 66 | ||
| XM_017353106.2 | XP_017208595.1 | 11 - 12 | 2684 | 66 | ||
| XM_021469872.1 | XP_021325547.1 | 12 - 13 | 2726 | 66 | ||
| XM_021469911.1 | XP_021325586.1 | 11 - 12 | 2644 | 66 | ||
| XM_021469944.1 | XP_021325619.1 | 11 - 12 | 2458 | 66 | ||
| XM_021469965.1 | XP_021325640.1 | 11 - 12 | 2415 | 66 | ||
| XM_021469996.1 | XP_021325671.1 | 11 - 12 | 2246 | 66 | ||
| XM_021470020.1 | XP_021325695.1 | 11 - 12 | 2235 | 66 | ||
| GenBank mRNA | BC049429.1 | - | 7 - 8 | 1139 | 66 | |
More Information
Set Membership: |
|

gab1
Genomic Map