Assay Details
Gene Symbol: prkab1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328554.1 | NP_001315483.1 | 5 - 6 | 637 | 109 | |
NM_214713.2 | NP_999878.1 | 5 - 6 | 682 | 109 | ||
GenBank mRNA | BC066758.1 | - | 5 - 6 | 597 | 109 | |
EH456078.1 | - | 5 - 6 | 630 | 109 | ||
EH461046.1 | - | 3 - 4 | 762 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
