Assay Details
Gene Symbol: prkab1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328554.1 | NP_001315483.1 | 6 - 7 | 775 | 68 | |
NM_214713.2 | NP_999878.1 | 6 - 7 | 820 | 68 | ||
GenBank mRNA | BC066758.1 | - | 6 - 7 | 735 | 68 | |
EH456078.1 | - | 6 - 7 | 768 | 68 | ||
EH461046.1 | - | 2 - 3 | 624 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
