Assay Details
| Gene Symbol: rps8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214793.1 | NP_999958.1 | 2 - 3 | 138 | 64 | |
| GenBank mRNA | AY561509.1 | - | 2 - 3 | 138 | 64 | |
| BC076163.1 | - | 2 - 3 | 138 | 64 | ||
More Information
Set Membership: |
|

rps8a
Genomic Map