Assay Details
| Gene Symbol: immt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001401.1 | NP_001001401.1 | 8 - 9 | 1037 | 61 | |
| XM_005157191.3 | XP_005157248.1 | 7 - 8 | 977 | 61 | ||
| GenBank mRNA | BC057438.1 | - | 8 - 9 | 1037 | 61 | |
More Information
Set Membership: |
|

immt
Genomic Map