Assay Details
Gene Symbol: ptenb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001822.2 | NP_001001822.1 | 1 - 2 | 681 | 75 | |
GenBank mRNA | AY398670.1 | - | 1 - 2 | 91 | 75 | |
AY398671.1 | - | 1 - 2 | 91 | 75 | ||
BC093110.1 | - | 1 - 2 | 688 | 75 | ||
BC107962.1 | - | 1 - 2 | 681 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
