Assay Details
| Gene Symbol: ptenb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001822.2 | NP_001001822.1 | 4 - 5 | 863 | 142 | |
| GenBank mRNA | AY398670.1 | - | 4 - 5 | 273 | 142 | |
| AY398671.1 | - | 4 - 5 | 273 | 142 | ||
| BC093110.1 | - | 4 - 5 | 870 | 142 | ||
| BC107962.1 | - | 4 - 5 | 863 | 142 | ||
More Information
Set Membership: |
|

ptenb
Genomic Map