Assay Details
| Gene Symbol: enoph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002226.1 | NP_001002226.1 | 2 - 3 | 312 | 72 | |
| GenBank mRNA | BC074060.1 | - | 2 - 3 | 312 | 72 | |
| BC164596.1 | - | 2 - 3 | 218 | 72 | ||
More Information
Set Membership: |
|

enoph1
Genomic Map