Assay Details
| Gene Symbol: kat5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002365.1 | NP_001002365.1 | 8 - 9 | 685 | 68 | |
| XM_009301297.3 | XP_009299572.1 | 7 - 8 | 684 | 68 | ||
| GenBank mRNA | BC076045.1 | - | 8 - 9 | 685 | 68 | |
More Information
Set Membership: |
|

kat5a
Genomic Map