Assay Details
| Gene Symbol: slc1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002666.1 | NP_001002666.2 | 1 - 2 | 213 | 63 | |
| XM_021477460.1 | XP_021333135.1 | 1 - 2 | 352 | 63 | ||
| GenBank mRNA | BC076553.1 | - | 1 - 2 | 213 | 63 | |
| BC163973.1 | - | 1 - 2 | 127 | 63 | ||
| HM138690.1 | - | 1 - 2 | 99 | 63 | ||
More Information
Set Membership: |
|

slc1a1
Genomic Map