Assay Details
| Gene Symbol: polr2eb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003564.2 | NP_001003564.1 | 2 - 3 | 325 | 74 | |
| GenBank mRNA | BC077151.1 | - | 2 - 3 | 329 | 74 | |
More Information
Set Membership: |
|

polr2eb
Genomic Map