Assay Details
| Gene Symbol: polr2eb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003564.2 | NP_001003564.1 | 4 - 5 | 515 | 124 | |
| GenBank mRNA | BC077151.1 | - | 4 - 5 | 519 | 124 | |
More Information
Set Membership: |
|

polr2eb
Genomic Map