Assay Details
Gene Symbol: polr2eb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003564.2 | NP_001003564.1 | 5 - 6 | 579 | 97 | |
GenBank mRNA | BC077151.1 | - | 5 - 6 | 583 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
